missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALDH2 Polyclonal Antibody

Description
ALDH2 Polyclonal Antibody for Western Blot, ICC/IF
ALDH2 belongs to the aldehyde dehydrogenase family of proteins. Aldehyde dehydrogenase is the second enzyme of the major oxidative pathway of alcohol metabolism. Two major liver isoforms of this enzyme, cytosolic and mitochondrial, can be distinguished by their electrophoretic mobilities, kinetic properties, and subcellular localizations. Most Caucasians have two major isozymes, while approximately 50% of Asians have only the cytosolic isozyme, missing the mitochondrial isozyme. A remarkably higher frequency of acute alcohol intoxication among Asians than among Caucasians could be related to the absence of the mitochondrial isozyme.
Specifications
Specifications
| Antigen | ALDH2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | Aldh2 |
| Gene Accession No. | P05091, P11884, P47738 |
| Gene Alias | acetaldehyde dehydrogenase 2; Ahd1; Ahd-1; Ahd5; Ahd-5; AHD-M1; aldehyde dehydrogenase 2 family (mitochondrial); aldehyde dehydrogenase 2, mitochondrial; aldehyde dehydrogenase, mitochondrial; ALDH class 2; ALDH1; ALDH2; ALDH-E2; ALDHI; ALDM; EC 1.2.1.3; liver mitochondrial ALDH; MGC1806; mitochondrial aldehyde dehydrogenase; nucleus-encoded mitochondrial aldehyde dehydrogenase 2 |
| Gene Symbols | Aldh2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?