missing translation for 'onlineSavingsMsg'
Learn More

AG126, MedChemExpress

Product Code. 16496897
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496897 5 mg 5mg
16435146 100 mg 100mg
16424188 10 mg 10mg
16422986 25 mg 25mg
16420537 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16496897 Supplier Medchem Express Supplier No. HY108330

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AG126 is a tyrosine kinase inhibitor which can prevent the activation of mitogen-activated protein kinase p42MAPK (ERK2).

AG126 is a tyrosine kinase inhibitor which can prevent the activation of mitogen-activated protein kinase p42MAPK (ERK2).

Specifications

Chemical Name or Material AG126
CAS 118409-62-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C10H5N3O3
Quantity 5 mg
Synonym Tyrphostin AG126
Solubility Information DMSO : 100 mg/mL (464.75 mM; Need ultrasonic and warming)
SMILES N#C/C(C#N)=C/C1=CC=C([N+]([O-])=O)C(O)=C1
Molecular Weight (g/mol) 215.17
Formula Weight 215.17
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.