missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenylate Cyclase 1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92583-0.02ml
This item is not returnable.
View return policy
Description
Adenylate Cyclase 1 Polyclonal antibody specifically detects Adenylate Cyclase 1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| Adenylate Cyclase 1 | |
| Polyclonal | |
| Western Blot 1:200-1:1000 | |
| 3'-5'-cyclic AMP synthetase, AC1, adenyl cyclase, adenylate cyclase 1 (brain), adenylate cyclase type 1, Adenylate cyclase type I, Adenylyl cyclase 1, ATP pyrophosphate-lyase 1, Ca(2+)/calmodulin-activated adenylyl cyclase, EC 4.6.1.1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 470-610 of human ADCY1 (NP_066939.1). NIETFFIVPSHRRKIFPGLILSDIKPAKRMKFKTVCYLLVQLMHCRKMFKAEIPFSNVMTCEDDDKRRALRTASEKLRNRSSFSTNVVYTTPGTRVNRYISRLLEARQTELEMADLNFFTLKYKHVEREQKYHQLQDEYFT | |
| 0.02 mL | |
| Lipid and Metabolism | |
| 107 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction