Learn More
Lenz Laborglasintrumente Adapters Straight, for Distilling Receivers acc. to Bredt
Description
- Straight adapters
- With two NS sockets
- Vacuum connection: GL 18
- Made of DURAN™ tubing
Specifications
Specifications
| Material | Duran Glass |
| Standard Taper Joint | Upper socket NS 29/32, Lower socket NS 29/32 |