missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Adapters with NS Stopcock, Bent

Product Code. 10264481
Change view
Click to view available options
Standard Taper Joint No.:
NS 19/26
NS 24/29
NS 29/32
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No. unitSize
10264481 NS 29/32 Each
10090481 NS 19/26 Each
10396021 NS 24/29 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10264481 Supplier Lenz Laborglasintrumente Supplier No. 05105004
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

  • Adapters with NS socket
  • Bent glass olive (90°) and NS stopcock with solid glass plug
  • Bore: 2,5 mm
  • Made of DURAN™ tubing

Specifications

Includes Bent glass olive (90°)
Material Borosilicate Glass (3.3)
Standard Taper Joint No. NS 29/32
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.