Learn More
Dostmann Electronic Adapter Plug
Description
- Adapter DIN connector to type K miniature connector
- Adapter for connecting type K probes with miniature connector on P600/T900-series
- Dimensions:100 mm cable length
- Weight: 25 g
Specifications
Specifications
| Probe Type | Type K Thermocouple |
| Product Type | Adapter Plug |