Learn More
Lenz Laborglasintrumente Adapter Duran DIN 12249 Socket
Product Code. 10189260
Change view
Click to view available options
Standard Taper Joint No:
NS 10/19
NS 12/21
NS 14/23
NS 19/26
NS 25/29
Unit Size:
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No unitSize
10189260 NS 14/23 Pack of 10
10625231 NS 10/19 Pack of 10
10772522 NS 12/21 Pack of 10
10322901 NS 25/29 Pack of 10
10414061 NS 19/26 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10189260 Supplier Lenz Laborglasintrumente Supplier No. 01100114
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Sockets
  • Acc. to DIN 12249 form E
  • DIN standard
  • Conical ground sockets
  • Made of DURAN™ tubing
Specifications
Standard Taper Joint No NS 14/23
Thickness (Metric) Wall 1.8 mm
Diameter (Metric) Outer Tube 17 mm
Material Duran Glass