missing translation for 'onlineSavingsMsg'
Learn More

Socorex™ Acura™ Electro 936 Macropipette

Product Code. 10050992
Change view
Click to view available options
Volume (Metric):
0.25 to 5 mL
0.5 to 10 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10050992 0.5 to 10 mL Each
11889223 0.25 to 5 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10050992 Supplier Socorex™ Supplier No. 936.10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ergonomically friendly, this electronic single-channel macropipette comes with a QC certificate, Qualitips™ pipette tip samples and operating instructions. Socorex™ Acura™ Electro 936 Macropipette charges quickly and has a long working autonomy.

  • Exceptional, user-friendly ergonomics.
  • Lightweight.
  • Easy, intuitive, self-teaching program.
  • Large display, reversible right/left reading.
  • Microprocessor-controlled motor.
  • Swift-interchange, rapidly charging, long-working, rechargeable battery pack.
  • Interchangeable nozzle protection filter.
  • Adapters for Pasteur pipette and straw tips.
  • Outstanding accuracy and precision.

Specifications

Accuracy <±1.0% - <±0.5%
Battery Type 4.8V NiMH
Certifications/Compliance EN ISO 8655, GMP, GLP, NCCLS. IVD 98/79 EEC (in vitro diagnostic medical devices), ED 89/336 EEC, 91/263 EEC, 92/31 EEC, 93/68 EEC, FCCCFR47, part 15 (electromagnetic compatibility), ED 73/23 EEC, 93/68EEC, IEC 61010-1 (electrical safety), ED 2011/65/EC Restriction of Hazardous Substances (RoHS)
Model 936
Number of Channels 1
Product Line Acura
Type Electronic Macropipette
Volume (Metric) 0.5 to 10 mL
Display Right/Left LCD
Includes QC certificate, Qualitips™ pipette tip samples and operating instructions.
Increments 0.05 mL
Precision <0.5% - <0.15%
Quantity 1 Each
Recharge Time <1.5 Hours
Warranty 24 Months
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.