missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid, potassium salt, 99+%, for analysis

Product Code. 10177303 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10177303 5 kg Plastic drum 5kg
10338640 500 g Plastic bottle 500g
10072830 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10177303 Supplier Thermo Scientific Acros Supplier No. 217100050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 127-08-2 | C2H3KO2 | 98.142 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]

Specifications

pH 7.5 to 8.5 (5% soln. at 20°C)
Melting Point 292.0°C
CAS Min % 99.0
CAS Max % 100.0
Color White
Assay Percent Range 99+%
Linear Formula CH3CO2K
Beilstein 02, 96
Fieser 01,906; 06,475
Merck Index 15, 7726
Solubility Information Solubility in water: 2694g/L (25°C). Other solubilities: soluble in alcohol, insoluble in ether
Formula Weight 98.14
Percent Purity ≥99%
Grade Analytical
Quantity 5 kg
Flash Point >250°C
Infrared Spectrum Authentic
Packaging Plastic drum
Physical Form Granular Powder
Chemical Name or Material Acetic acid, potassium salt, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.