missing translation for 'onlineSavingsMsg'
Learn More

Acetic acid glacial 99-100%, BAKER, J.T. Baker™

Product Code. 10127950 Shop All J.T.Baker Products
Change view
Click to view available options
:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10127950 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10127950 Supplier JT Baker Supplier No. 6003.2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Meets Reagent Specifications for testing Ph. Eur. Monographs

Specifications

CAS 64-19-7
Assay Percent Range 99.6 to 100.5 %
Molecular Formula C2H4O2
MDL Number MFCD00036152
Synonym Ethanoic acid
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
SMILES CC(O)=O
IUPAC Name acetic acid
Molecular Weight (g/mol) 60.05
Quantity 2.5 L
Packaging Plastic bottle
Residue after Evaporation max. 0.005 %
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.