missing translation for 'onlineSavingsMsg'
Learn More

Acetazolamide D3 (methyl D3), Dr. Ehrenstorfer

Product Code. 30308923
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
30308923 10 mg 10mg
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30308923

Brand: Dr. Ehrenstorfer DREC10017810

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetazolamide-d3
CAS 1189904-01-5
Molecular Formula C4H3D3N4O3S2
Quantity 10 mg
Synonym N-[5-Aminosulfonyl)-1,3,4-thiadiazol-2-yl]acetamide-d3, 5-Acetamido-1,3,4-thiadiazole-2-sulfonamide-d3, Acetamox-d3, Atenezol-d3, Defiltran-d3, Diamox-d3, Didoc-d3, Acetamide-2,2,2-d3, N-[5-(aminosulfonyl)-1,3,4-thiadiazol-2-yl]-, N-[5-(Aminosulfonyl)-1,3,4-thiadiazol-2-yl]acetamide-2,2,2-d3, 2,2,2-Trideuterio-N-(5-sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide, 2-(Acetylamino-d3)-1,3,4-thiadiazole-5-sulfonamide, 5-(Acetamido-d3)-1,3,4-thiadiazole-2-sulfonamide, Acetazolamide-D3 (methyl-D3), Acetazolamide D3 (methyl D3)
InChI Formula InChI=1S/C4H6N4O3S2/c1-2(9)6-3-7-8-4(12-3)13(5,10)11/h1H3,(H2,5,10,11)(H,6,7,9)/i1D3
SMILES [2H]C([2H])([2H])C(=O)Nc1nnc(s1)S(=O)(=O)N
IUPAC Name 2,2,2-trideuterio-N-(5-sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide
Molecular Weight (g/mol) 225.26
Formula Weight 225.007 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.