missing translation for 'onlineSavingsMsg'
Learn More

Acetaldehyde, 99%

Product Code. p-7050009
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 x 500 mL
Unit Size:
100mL
2000mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387548 100 mL 100mL
11484980 500 mL 500mL
11494980 4 x 500 mL 2000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11387548 Supplier Thermo Scientific Alfa Aesar Supplier No. 033244.AE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 75-07-0 | C2H4O | 44.053 g/mol

Acetaldehyde is used in the production of drugs, perfumes, explosives, lacquers and varnishes, photographic chemicals, phenolic and urea resins, rubber accelerators and antioxidants. It is used as an intermediate for the synthesis of acetic acid, ethyl acetate, acetic anhydride, pentaerythritol, peracetic acid, chloral and glyoxal. As a solvent, it is used in rubber production, tanning of leather and in the paper industry. It is used as a fruit and fish preservative, as a flavoring agent, as a denaturant for alcohol and in composition of fuel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-07-0
Molecular Formula C2H4O
Molecular Weight (g/mol) 44.053
MDL Number MFCD00006991
InChI Key IKHGUXGNUITLKF-UHFFFAOYSA-N
Synonym ethanal, acetic aldehyde, ethyl aldehyde, acetaldehyd, acetylaldehyde, aldehyde, acetic ethanol, aldeide acetica, octowy aldehyd, aldehyde acetique
PubChem CID 177
ChEBI CHEBI:15343
IUPAC Name acetaldehyde
SMILES CC=O

Specifications

Melting Point -128°C
Density 0.780 g/mL
Boiling Point 20.8°C
Flash Point -27°C (-17°F)
Refractive Index 1.3316
Quantity 100 mL
UN Number 1089
Beilstein 505984
Sensitivity Air Sensitive
Merck Index 14,39
Solubility Information Miscible with water,ethanol,ether,benzene,toluene,xylene,turpentine,acetone and gasoline.Slightly miscible with chloroform.
Formula Weight 44.06
Percent Purity 98.5%
Physical Form Liquid
Chemical Name or Material Acetaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.