missing translation for 'onlineSavingsMsg'
Learn More

Abemaciclib, MedChemExpress

Product Code. p-200101572
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491216 200 mg 200mg
16479567 5 mg 5mg
16441806 500 mg 500mg
16433077 50 mg 50mg
16427086 100 mg 100mg
16417028 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16491216 Supplier Medchem Express Supplier No. HY16297A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Abemaciclib (LY2835219) is a selective CDK4/6 inhibitor with IC50 values of 2 nM and 10 nM for CDK4 and CDK6, respectively.

Abemaciclib (LY2835219) is a selective CDK4/6 inhibitor with IC50 values of 2 nM and 10 nM for CDK4 and CDK6, respectively.

Specifications

Chemical Name or Material Abemaciclib
CAS 1231929-97-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H32F2N8
Quantity 200 mg
Synonym LY2835219
Solubility Information DMSO : 5 mg/mL (9.87 mM; Need ultrasonic)
SMILES CC1=NC2=C(F)C=C(C3=NC(NC4=NC=C(CN5CCN(CC)CC5)C=C4)=NC=C3F)C=C2N1C(C)C
Molecular Weight (g/mol) 506.59
Formula Weight 506.59
Percent Purity 99.06%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.