missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ A2B5 Monoclonal Antibody (105)
GREENER_CHOICE

Product Code. 10124883
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10124883 100 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10124883 Supplier Invitrogen™ Supplier No. 433110

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Mouse Monoclonal Antibody

A2B5 Monoclonal Antibody for Western Blot, ICC/IF, IHC, Flow

A2B5 binds to neural tissue such as retina, brain, spinal cord, and dorsal root ganglia. It is also able to bind plasma membranes of human, rat, chicken and mouse islet cells and can be used in cell sorting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen A2B5
Applications Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 105
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene Alias A2B5 marker
Host Species Mouse
Immunogen Chicken embryo retina cells.
Purification Method IgM
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Chicken
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.