missing translation for 'onlineSavingsMsg'
Learn More
Learn More
A-967079, MedChemExpress
Description
A-967079 is a selective TRPA1 receptor antagonist with IC50s of 67 nM and 289 nM at human and rat TRPA1 receptors, respectively, and has good penetration into the CNS.
Specifications
Specifications
| Chemical Name or Material | A-967079 |
| CAS | 1170613-55-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C12H14FNO |
| Quantity | 25 mg |
| Solubility Information | DMSO : 100 mg/mL (482.53 mM; Need ultrasonic) |
| SMILES | CCC(/C(C)=C/C1=CC=C(F)C=C1)=N\O.[E].[E] |
| Molecular Weight (g/mol) | 207.24 |
| Formula Weight | 207.24 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?