missing translation for 'onlineSavingsMsg'
Learn More

7-Methoxyindole, 97%, Thermo Scientific™

Product Code. 10001284 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10001284 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10001284 Supplier Thermo Scientific Acros Supplier No. 397520010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 3189-22-8
CAS Min % 96.0
CAS Max % 100.0
Color Brown to Yellow
Density 1.1260g/mL
Boiling Point 166°C (27.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C9H9NO
MDL Number MFCD00047203
Quantity 1 g
Specific Gravity 1.126
Synonym 7-methoxyindole, 7-methoxy indole, 1h-indole, 7-methoxy, indole, 7-methoxy, methoxyindole 7-, unii-9r05qk9rp4, 7-methoxy-indole, pubchem7434, acmc-209hpi
InChI Key FSOPPXYMWZOKRM-UHFFFAOYSA-N
SMILES COC1=CC=CC2=C1NC=C2
IUPAC Name 7-methoxy-1H-indole
Molecular Weight (g/mol) 147.18
PubChem CID 76660
Formula Weight 147.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 7-Methoxyindole
Show More Show Less
compliance-icons
Product Identifier
  • 7-Methoxyindole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.