missing translation for 'onlineSavingsMsg'
Learn More

(6E)-7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3-hydroxy-5-oxohept-6-enoic Acid (5-Ketofluvastatin), Mikromol™

Product Code. 17971616
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17971616 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17971616 Supplier Mikromol Supplier No. MM0676.010025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(6E)-7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3-hydroxy-5-oxohept-6-enoic Acid (5-Ketofluvastatin)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Ketofluvastatin
CAS 1160169-39-0
Analyte or Component Names 5-Ketofluvastatin
Shipping Condition Room Temperature
Molecular Formula C24H24FNO4
Certifications/Compliance ISO 17025
Synonym (6E)-7-[3-(4-Fluorophenyl)-1-(1-methylethyl)-1H-indol-2-yl]-3-hydroxy-5-oxo-6-heptenoic Acid, 5-Ketofluvastatin, Fluvastatin Sodium Impurity D, Fluvastatin Impurity D
InChI Formula InChI=1S/C24H24FNO4/c1-15(2)26-21-6-4-3-5-20(21)24(16-7-9-17(25)10-8-16)22(26)12-11-18(27)13-19(28)14-23(29)30/h3-12,15,19,28H,13-14H2,1-2H3,(H,29,30)/b12-11+
SMILES CC(C)n1c(\C=C\C(=O)CC(O)CC(=O)O)c(c2ccc(F)cc2)c3ccccc13
IUPAC Name (E)-7-[3-(4-fluorophenyl)-1-isopropyl-indol-2-yl]-3-hydroxy-5-oxo-hept-6-enoic acid
Molecular Weight (g/mol) 409.45
Quantity 25 mg
Formula Weight 409.1689 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.