missing translation for 'onlineSavingsMsg'
Learn More

6alpha,9-Difluoro-17-[[(fluoromethyl)sulphanyl]carbonyl]-16alpha-methyl-3,11-dioxo-androsta-1,4-dien-17alpha-yl Propanoate, Mikromol™

Product Code. 17921286
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17921286 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17921286 Supplier Mikromol Supplier No. MM1244.060025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

6alpha,9-Difluoro-17-[[(fluoromethyl)sulphanyl]carbonyl]-16alpha-methyl-3,11-dioxo-androsta-1,4-dien-17alpha-yl Propanoate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 11-Oxo Fluticasone Propionate
CAS 1219174-94-3
Analyte or Component Names 11-Oxo Fluticasone Propionate
Shipping Condition Room Temperature
Molecular Formula C25 H29 F3 O5 S
Certifications/Compliance ISO 9001
Synonym Androsta-1,4-diene-17-carbothioic acid, 6,9-difluoro-16-methyl-3,11-dioxo-17-(1-oxopropoxy)-, S-(fluoromethyl) ester, (6α,16α,17α)-, 6α,9-difluoro-17-[[(fluoromethyl)sulfanyl]carbonyl]-16α-methyl-3,11-dioxoandrosta-1,4-dien-17α-yl propanoate, Fluticasone Propionate Imp. F (EP), GR 40775X, Ph Eur Fluticasone Propionate Impurity F, 11-keto fluticasone propionate (GR 40775X), unidentified impurity with RRT 1.23, disulphide dipropionate dithioketone (GR 247095X), dithio (GR 269949X), Compound A (fluticasone propionate related)
InChI Formula InChI=1S/C25H29F3O5S/c1-5-20(31)33-25(21(32)34-12-26)13(2)8-15-16-10-18(27)17-9-14(29)6-7-22(17,3)24(16,28)19(30)11-23(15,25)4/h6-7,9,13,15-16,18H,5,8,10-12H2,1-4H3/t13-,15+,16+,18+,22+,23+,24+,25+/m1/s1
SMILES CCC(=O)O[C@@]1([C@H](C)C[C@H]2[C@@H]3C[C@H](F)C4=CC(=O)C=C[C@]4(C)[C@@]3(F)C(=O)C[C@]12C)C(=O)SCF
IUPAC Name [(6S,8S,9R,10S,13S,14S,16R,17R)-6,9-difluoro-17-(fluoromethylsulfanylcarbonyl)-10,13,16-trimethyl-3,11-dioxo-7,8,12,14,15,16-hexahydro-6H-cyclopenta[a]phenanthren-17-yl] propanoate
Molecular Weight (g/mol) 498.56
Quantity 25 mg
Formula Weight 498.1688 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.