Learn More
6-Heptenoic acid, 96%
Description
6-Heptenoic acid is used in the preparation of 6-(iodomethyl)-hexanolide through iodo-lactonization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
6-Heptenoic acid is used in the preparation of 6-(iodomethyl)-hexanolide through iodo-lactonization.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents, strong acids and bases.
Specifications
Specifications
| Melting Point | ∼-6°C |
| Density | 0.946 |
| Boiling Point | 222°C to 224°C |
| Flash Point | >110°C (230°F) |
| Odor | Unpleasant |
| Refractive Index | 1.439 |
| Quantity | 1 g |
| UN Number | UN3265 |
| Beilstein | 1747265 |
| Solubility Information | Miscible with water. |
| Show More |
Safety and Handling
- 6-Heptenoic acid
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: Hept-6-enoic acid
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.