missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Mueller Hinton Agar (Dehydrated)
GREENER_CHOICE

Product Code. 10096002
Change view
Click to view available options
Quantity:
500 g
2.5kg
5 kg
Unit Size:
2.5kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10096002 5 kg 5kg
10452453 500 g 500g
12771500 2.5kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10096002 Supplier Thermo Scientific™ Supplier No. CM0337T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Perform effortless Antimicrobial Susceptibility Testing (AST) while conforming to EUCAST and CLSI with this medium.

Perform effortless Antimicrobial Susceptibility Testing (AST) using Thermo Scientific™ Oxoid™ Mueller Hinton Agar (Dehydrated), a medium designed and manufactured according to the European standard, EUCAST1 and the American standard, CLSI2, 3, 4.

The major use of Mueller-Hinton Agar is for Antimicrobial Susceptibility Testing (AST). It has become the standard medium for the Kirby-Bauer method5, 6 and can be used in internationally recognized standard procedures.

  • Specified by the Clinical and Laboratory Standards Institute (CLSI) formerly the National Committee for Clinical Laboratory Standards (NCCLS) 2,3,4 and The European Committee on Antimicrobial Susceptibility Testing (EUCAST)1
  • Standard medium for the Kirby-Bauer method5, 6

The inclusion of starch in the medium ensures that toxic factors produced during microbial growth will be absorbed, and its presence is often essential to establish growth from very small inocula.

Antibiotic susceptibility tests are performed in accordance with and meet the acceptance limits of the current ISO/TS 16782 using the following microorganisms:

Staphylococcus aureus ATCC®25923 WDCM00034

Staphylococcus aureus ATCC®29213 WDCM00131

Staphylococcus aureus ATCC®43300 WDCM00211

Staphylococcus aureus NCTC 12493 WDCM00212 Escherichia coli ATCC®25922 WDCM00013

Escherichia coli ATCC®35218

Pseudomonas aeruginosa ATCC®27853 WDCM00025

Enterococcus faecalis ATCC®33186 WDCM00210 Enterococcus faecalis ATCC®29212 WDCM00087

Streptococcus pneumoniae ATCC®49619

Haemophilus influenzae ATCC®49247

Haemophilus influenzae ATCC®49766

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Conforms to CLSI standards. Industrial Reference: FDA.
Form Powder
Product Type Agar
Quantity 5 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.