missing translation for 'onlineSavingsMsg'
Learn More

5-Bromopyridine-3-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. p-3541030
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10546894 250 mg Amber Glass Bottle 250mg
10779015 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10546894 Supplier Thermo Scientific Maybridge Supplier No. CC67103CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 65001-21-0
Molecular Formula C5H3BrClNO2S
Molecular Weight (g/mol) 256.498
MDL Number MFCD08235263
InChI Key AVILRJQPDYPXFQ-UHFFFAOYSA-N
Synonym 5-bromopyridine-3-sulphonyl chloride, 5-bromo-3-pyridinesulfonyl chloride, 5-bromopyridine-3-sulfonylchloride, 5-bromo-pyridine-3-sulfonyl chloride, 5-bromo 3-pyridyl chlorosulfone, 5-bromopyridin-3-sulfonyl chloride, 3-bromopyridine-5-sulfonyl chloride, 5-bromopyridine-3-sulphonylchloride, 5-bromo-3-chlorosulphonyl pyridine, 3-pyridinesulfonylchloride, 5-bromo
PubChem CID 20324474
IUPAC Name 5-bromopyridine-3-sulfonyl chloride
SMILES C1=C(C=NC=C1Br)S(=O)(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 256.51
Percent Purity 97%
Chemical Name or Material 5-Bromopyridine-3-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.