missing translation for 'onlineSavingsMsg'
Learn More

5-Acetamidonaphthalene-1-sulfonamide, TRC

Product Code. 30309342
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30309342 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309342 Supplier Toronto Research Chemicals Supplier No. TRCA1584601G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Acetamidonaphthalene-1-sulfonamide
CAS 32327-48-3
Molecular Formula C12 H12 N2 O3 S
Quantity 1 g
InChI Formula InChI=1S/C12H12N2O3S/c1-8(15)14-11-6-2-5-10-9(11)4-3-7-12(10)18(13,16)17/h2-7H,1H3,(H,14,15)(H2,13,16,17)
SMILES CC(=O)Nc1cccc2c(cccc12)S(=O)(=O)N
IUPAC Name N-(5-sulfamoylnaphthalen-1-yl)acetamide
Molecular Weight (g/mol) 264.3
Formula Weight 264.0569
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.