missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dichloro-1-indanone, 97%

Product Code. 15440448
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15440448 1 g 1g
15450448 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15440448 Supplier Thermo Scientific Alfa Aesar Supplier No. H54007.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 448193-94-0 | C9H6Cl2O | 201.05 g/mol

5,7-Dichloro-1-indanone is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5,7-Dichloro-1-indanone is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 448193-94-0
Molecular Formula C9H6Cl2O
Molecular Weight (g/mol) 201.05
MDL Number MFCD09908160
InChI Key TWNXYJNWNZZECV-UHFFFAOYSA-N
Synonym 5,7-dichloro-2,3-dihydro-1h-inden-1-one, 5,7-dichloro-1-indanone, 1h-inden-1-one, 5,7-dichloro-2,3-dihydro, 5,7-dichloroindan-1-one, acmc-1asf2
PubChem CID 22292346
SMILES ClC1=CC(Cl)=C2C(=O)CCC2=C1

Specifications

Melting Point 98°C
Quantity 1 g
Solubility Information Slightly soluble in water.
IUPAC Name 5,7-dichloro-2,3-dihydro-1H-inden-1-one
Formula Weight 201.05
Percent Purity 97%
Chemical Name or Material 5,7-Dichloro-1-indanone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.