missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BRAND™ PCR products deliver excellent, reproducible results for a wide range of PCR and qPCR applications. Manufactured from high-quality virgin resins, these products are free from lubricants, chemical additives, DNA, DNase, RNase, and other PCR inhibitors. Key features include:
- Exceptional Quality and Purity
- BIO-CERT™ PCR QUALITY ensures plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors
- Free of contaminants that could interfere with reagents and samples
- Ultra-smooth vessel interiors minimize enzyme nucleic acid binding
- Precision Thermal Performance
- Thin, uniform walls ensure consistent, precise thermal transfer for reliable results
- Designed for rapid and accurate heat transfer, reducing cycle times
- Wide Compatibility
- Compact design is compatible with most standard thermal cyclers, ensuring convenient handling
- White PCR plates, enhanced with titanium dioxide (TiO2), optimize fluorescence signal reflection and are recommended for qPCR applications
- User-Friendly Features
- Blue alphanumeric coding for easy sample identification
- Well rims designed to prevent cross-contamination and ensure reliable sealing with tailored mats
Specifications
Specifications
| Barcode | No Barcode |
| Binding Property | Untreated |
| Certifications/Compliance | RNase, DNase, DNA-free |
| Format | Standard |
| Array | 6 x 8 |
| Autoclavable | Autoclavable |
| Color | White |
| Disposable | Single-use |
| Product Type | qPCR 48-well plate |
| For Use With (Equipment) | Thermal Cyclers |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
