missing translation for 'onlineSavingsMsg'
Learn More

RETSCH 400 dia. x 65 mmH Stainless Steel Test Sieve, ISO Certified

Product Code. 10064921 Shop All Retsch Products
Change view
Click to view available options
Pore Size:
25 μm
32 μm
36 μm
38 μm
40 μm
45 μm
50 μm
53 μm
56 μm
63 μm
71 μm
75 μm
80 μm
90 μm
100 μm
106 μm
112 μm
125 μm
140 μm
150 μm
160 μm
180 μm
200 μm
212 μm
224 μm
250 μm
280 μm
300 μm
315 μm
355 μm
400 μm
425 μm
450 μm
500 μm
560 μm
600 μm
630 μm
710 μm
800 μm
850 μm
900 μm
71 mm
75 mm
80 mm
90 mm
100 mm
106 mm
112 mm
125 mm
Unit Size:
Each
49 product options available for selection
Product selection table with 49 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size unitSize
10064921 250 μm Each
10781623 100 mm Each
10514882 106 mm Each
10145981 100 μm Each
10370862 112 mm Each
10488921 106 μm Each
10546222 112 μm Each
10750083 125 mm Each
10620492 125 μm Each
10187021 140 μm Each
10630462 150 μm Each
10600292 160 μm Each
10248691 180 μm Each
10166791 200 μm Each
10486811 212 μm Each
10268831 224 μm Each
10791223 25 μm Each
10670462 280 μm Each
10447201 315 μm Each
10419661 32 μm Each
10390812 300 μm Each
10300822 355 μm Each
10525252 36 μm Each
10258581 38 μm Each
10360862 400 μm Each
10721613 40 μm Each
10749892 425 μm Each
10258731 450 μm Each
10136591 45 μm Each
10390242 500 μm Each
10369601 50 μm Each
10218681 53 μm Each
10593332 560 μm Each
10761813 56 μm Each
10740083 600 μm Each
10563912 630 μm Each
10438351 63 μm Each
10268921 710 μm Each
10711053 71 mm Each
10749322 71 μm Each
10167071 75 mm Each
10516032 75 μm Each
10700473 800 μm Each
10034981 80 μm Each
10105981 850 μm Each
10340722 80 mm Each
10668571 900 μm Each
10147171 90 mm Each
10670282 90 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 49 options available.
49 options
This item is not returnable. View return policy
Product Code. 10064921 Supplier RETSCH Supplier No. 60.166.000250
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Quantity 1Each
Pore Size 250 μm
Type Test Sieve
Certifications/Compliance ISO 3310/1
Material Stainless Steel
For Use With (Equipment) Shaker AS 450
Height (Metric) 65mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 400 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.