missing translation for 'onlineSavingsMsg'
Learn More

[4-(tert-Butoxycarbonyl)piperazin-1-yl]acetic acid, 97%, Thermo Scientific™

Product Code. 10071054
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10071054 5 g Amber Glass Bottle 5g
10142844 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10071054 Supplier Thermo Scientific Maybridge Supplier No. MO08453DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 156478-71-6
Molecular Formula C11H20N2O4
Molecular Weight (g/mol) 244.291
MDL Number MFCD02682402
InChI Key WZBHMXRBXXCEDD-UHFFFAOYSA-N
Synonym 2-4-tert-butoxycarbonyl piperazin-1-yl acetic acid, 4-boc-1-piperazineacetic acid, 1-boc-4-carboxymethyl piperazine, 4-tert-butoxycarbonyl piperazin-1-yl acetic acid, 4-boc-piperazineacetic acid, 2-1-boc-piperazin-4-yl-acetic acid, boc-4-carboxymethyl piperazine, 1-piperazineacetic acid, 4-1,1-dimethylethoxy carbonyl, 2-4-tert-butoxy carbonyl piperazin-1-yl acetic acid, 4-carboxymethyl-piperazine-1-carboxylic acid tert-butyl ester
PubChem CID 2735642
IUPAC Name 2-[4-[(2-methylpropan-2-yl)oxycarbonyl]piperazin-1-yl]acetic acid
SMILES CC(C)(C)OC(=O)N1CCN(CC1)CC(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 244.29
Percent Purity 97%
Chemical Name or Material [4-(Tert-butoxycarbonyl)piperazin-1-yl]acetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.