missing translation for 'onlineSavingsMsg'
Learn More

4-Pyridineacetonitrile, ≥97%, Thermo Scientific Chemicals

Product Code. p-7055887
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15441869 250 mg 250mg
15431869 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15441869 Supplier Thermo Scientific Alfa Aesar Supplier No. H64588.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A pyridine derivative used in the preparation of inhibitors of Acyl CoA, cholesterol acyltransferase as well as bone morphogenetic protein (BMP) signaling inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A pyridine derivative used in the preparation of inhibitors of Acyl CoA, cholesterol acyltransferase as well as bone morphogenetic protein (BMP) signaling inhibitors.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13121-99-8
Molecular Formula C7H6N2
Molecular Weight (g/mol) 118.14
MDL Number MFCD03412601
InChI Key BMVSAKPRNWZCPG-UHFFFAOYSA-N
Synonym pyridin-4-yl-acetonitrile, 2-pyridin-4-yl acetonitrile, 4-pyridylacetonitrile, 4-pyridineacetonitrile, pyridin-4-ylacetonitrile, pyridine-4-acetonitrile, rarechem ak ml 0204, 2-4-pyridyl acetonitrile, 4-pyridylacetonitrilehydrochloride, 4-cyanomethyl-pyridine
PubChem CID 4112085
IUPAC Name 2-pyridin-4-ylacetonitrile
SMILES N#CCC1=CC=NC=C1

Specifications

Quantity 250 mg
UN Number UN3439
Beilstein 471559
Solubility Information Slightly soluble in water.
Formula Weight 118.14
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material 4-Pyridineacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.