missing translation for 'onlineSavingsMsg'
Learn More

4-Piperidone monohydrate hydrochloride, 98%

Product Code. 10083613 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10083613 250 g Glass bottle 250g
10722591 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10083613 Supplier Thermo Scientific Acros Supplier No. 166192500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 40064-34-4 | HCl·H2O | 153.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40064-34-4
Molecular Formula HCl·H2O
Molecular Weight (g/mol) 153.61
MDL Number MFCD00012776
Synonym piperidine-4,4-diol hydrate hydrochloride

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 89°C to 99°C
Color Beige-Yellow to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 250 g
Beilstein 21, I, 262
Formula Weight 153.61
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 4-Piperidone monohydrate hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 4-Piperidone monohydrate hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.