missing translation for 'onlineSavingsMsg'
Learn More

4-Nonyl Phenol Monoethoxylate, TRC

Product Code. 30470829
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30470829 2.5 mg 2.5mg
30476310 5 mg 5mg
30469230 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30470829 Supplier Toronto Research Chemicals Supplier No. TRCN6504502.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-35-8
Molecular Formula C17 H28 O2
Molecular Weight (g/mol) 264.4
Synonym 4-Nonylphenol Ethoxylate, Ethanol, 2-(p-nonylphenoxy)- (7CI,8CI), 2-(4-Nonylphenoxy)ethanol, 2-(4-Nonylphenoxy)ethyl alcohol, 2-(p-Nonylphenoxy)ethanol, 4-Nonylphenol monoethoxylate, Ethylene glycol mono-p-nonylphenyl ether, Ethylene glycol p-nonylphenyl ether
IUPAC Name 2-(4-nonylphenoxy)ethanol
SMILES CCCCCCCCCc1ccc(OCCO)cc1

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C17H28O2/c1-2-3-4-5-6-7-8-9-16-10-12-17(13-11-16)19-15-14-18/h10-13,18H,2-9,14-15H2,1H3
Formula Weight 264.2089
Percent Purity >95%
Chemical Name or Material 4-Nonylphenol Monoethoxylate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.