missing translation for 'onlineSavingsMsg'
Learn More

(4-Methylpiperazino)(3-piperidinyl)methanone dihydrochloride, 97%, Thermo Scientific™

Product Code. 10063972
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10063972 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10063972 Supplier Thermo Scientific Maybridge Supplier No. MO01168DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 690632-07-6
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C11H23Cl2N3O
Packaging Amber Glass Bottle
MDL Number MFCD05865123
Synonym 4-methylpiperazino 3-piperidinyl methanone dihydrochloride, 1-methyl-4-piperidine-3-carbonyl piperazine dihydrochloride, 4-methylpiperazin-1-yl piperidin-3-yl methanone dihydrochloride, 4-methylpiperazin-1-yl-piperidin-3-ylmethanone dihydrochloride, 4-methyl-piperazin-1-yl-piperidin-3-yl-methanone dihydrochloride, 4-methylpiperazin-1-yl piperidin-3-yl methanone-hydrogen chloride 1/2
InChI Key RLSOTIZPTQYRRZ-UHFFFAOYSA-N
SMILES CN1CCN(CC1)C(=O)C2CCCNC2.Cl.Cl
IUPAC Name (4-methylpiperazin-1-yl)-piperidin-3-ylmethanone;dihydrochloride
Molecular Weight (g/mol) 284.225
Quantity 1 g
PubChem CID 2794686
Formula Weight 284.23
Percent Purity 97%
Chemical Name or Material (4-Methylpiperazino)(3-piperidinyl)methanone dihydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.