missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-3-oxo-N-phenylpentanamide, Mikromol

Product Code. 30460805
Change view
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30460805 4 x 25 mg -
17921886 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30460805 Supplier Mikromol Supplier No. MM0400.3200254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

4-Methyl-3-oxo-N-phenylpentanamide, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Phenyl Isobutyrylacetamide
CAS 124401-38-3
Analyte or Component Names N-Phenyl Isobutyrylacetamide
Shipping Condition Room Temperature
Molecular Formula C12 H15 N O2
Certifications/Compliance ISO 17025
Synonym 4-Methyl-3-oxo-N-phenylpentanamide, 4-Methyl-3-oxopentanoic acid anilide, 4-Methyl-3-oxopentanoic acid phenylamide, N-Phenyl-4-methyl-3-oxopentanamide, N-Phenyl Isobutyrylacetamide
InChI Formula InChI=1S/C12H15NO2/c1-9(2)11(14)8-12(15)13-10-6-4-3-5-7-10/h3-7,9H,8H2,1-2H3,(H,13,15)
Solution Type Neat
SMILES CC(C)C(=O)CC(=O)Nc1ccccc1
IUPAC Name 4-methyl-3-oxo-N-phenylpentanamide
Molecular Weight (g/mol) 205.25
Quantity 4 x 25 mg
Formula Weight 205.1103 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.