missing translation for 'onlineSavingsMsg'
Learn More

4-Isopropylbenzonitrile, 95%, Thermo Scientific™

Product Code. 10128264
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10128264 25 g 25g
10279924 5 g 5g
10406482 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10128264 Supplier Thermo Scientific Maybridge Supplier No. SB00932EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 13816-33-6
Molecular Formula C10H11N
Molecular Weight (g/mol) 145.205
MDL Number MFCD00052966
InChI Key YFDJCWXBKWRDPW-UHFFFAOYSA-N
Synonym 4-isopropylbenzonitrile, cuminyl nitrile, p-cyanocumene, benzonitrile, 4-1-methylethyl, cuminonitrile, cuminic nitrile, p-isopropylbenzonitrile, benzonitrile, p-isopropyl, 4-1-methylethyl benzonitrile, 4-propan-2-yl benzonitrile
PubChem CID 26289
IUPAC Name 4-propan-2-ylbenzonitrile
SMILES CC(C)C1=CC=C(C=C1)C#N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Linear Formula (CH3)2CHC6H4CN
Quantity 25 g
Formula Weight 145.2
Percent Purity 95%
Chemical Name or Material 4-Isopropylbenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.