missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2,5-dimethoxybenzaldehyde, 98+%

Product Code. 11471060
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11471060 5 g 5g
11481060 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11471060 Supplier Thermo Scientific Alfa Aesar Supplier No. B25500.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 31558-41-5 | C9H9BrO3 | 245.072 g/mol

4-Bromo-2,5-dimethoxybenzene is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-2,5-dimethoxybenzene is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions, in well sealed conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31558-41-5
Molecular Formula C9H9BrO3
Molecular Weight (g/mol) 245.072
MDL Number MFCD00156964
InChI Key BIFWGDWGCZLCHF-UHFFFAOYSA-N
Synonym 2,5-dimethoxy-4-bromobenzaldehyde, pubchem2068, acmc-1aiq9, cbmicro_047429, cambridge id 6238031, 4-bromo-2,5-dimethoxy benzaldehyde, 4-bromo-2,5-dimethoxy-benzaldehyde, 4-bromanyl-2,5-dimethoxy-benzaldehyde, benzaldehyde, 4-bromo-2,5-dimethoxy
PubChem CID 777997
IUPAC Name 4-bromo-2,5-dimethoxybenzaldehyde
SMILES COC1=CC(=C(C=C1C=O)OC)Br

Specifications

Melting Point 132°C to 135°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 245.08
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-2,5-dimethoxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.