missing translation for 'onlineSavingsMsg'
Learn More

4,4'-DDM, TRC

Product Code. 30366231
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366231 100 mg 100mg
30362824 500 mg 500mg
30349221 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30366231 Supplier Toronto Research Chemicals Supplier No. TRCD195303100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-76-8
Molecular Formula C13 H10 Cl2
Molecular Weight (g/mol) 237.12
Synonym Methane, bis(p-chlorophenyl)- (6CI,7CI,8CI), 1,1'-Methylenebis[4-chlorobenzene], 4,4'-Dichlorodiphenylmethane, Bis(4-chlorophenyl)methane, Bis(p-chlorophenyl)methane, DBM, DDM, NSC 406594, p,p'-DDM, p,p'-Dichlorodiphenylmethane
IUPAC Name 1-chloro-4-[(4-chlorophenyl)methyl]benzene
SMILES Clc1ccc(Cc2ccc(Cl)cc2)cc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H10Cl2/c14-12-5-1-10(2-6-12)9-11-3-7-13(15)8-4-11/h1-8H,9H2
Formula Weight 236.016
Chemical Name or Material 4,4'-DDM
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.