missing translation for 'onlineSavingsMsg'
Learn More

4-(2,3-Dihydro-1,4-benzodioxin-6-yl)-4-oxobutanoic acid, 97%, Thermo Scientific™

Product Code. 10079061
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10079061 10 g Amber Glass Bottle 10g
10592272 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10079061 Supplier Thermo Scientific Maybridge Supplier No. BTB12838EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 54557-81-2
Molecular Formula C12H12O5
Molecular Weight (g/mol) 236.223
MDL Number MFCD00168683
InChI Key LMDXEMFSAHAGGP-UHFFFAOYSA-N
Synonym 4-2,3-dihydro-1,4-benzodioxin-6-yl-4-oxobutanoic acid, 4-2,3-dihydro-benzo 1,4 dioxin-6-yl-4-oxo-butyric acid, 4-2,3-dihydrobenzo b 1,4 dioxin-6-yl-4-oxobutanoic acid, 4-3,4-ethylenedioxy phenyl-4-oxobutyric acid, 4-2,3-dihydro-1,4,benzodioxin-6-yl-4-oxobutanoic acid, 4-2,3-dihydrobenzo b 1,4 dioxin-7-yl-4-oxobutanoic acid, 4-oxo-4-2,3-dihydro-1,4-benzodioxin-6-yl butyric acid, 4-2h,3h-benzo 3,4-e 1,4-dioxin-6-yl-4-oxobutanoic acid, maybridge1_001974, regid_for_cid_2758835
PubChem CID 2758835
IUPAC Name 4-(2,3-dihydro-1,4-benzodioxin-6-yl)-4-oxobutanoic acid
SMILES C1COC2=C(O1)C=CC(=C2)C(=O)CCC(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 236.22
Percent Purity 97%
Chemical Name or Material 4-(2, 3-Dihydro-1, 4-benzodioxin-6-yl)-4-oxobutanoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.