missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Pyrazol-1-yl)benzaldehyde, ≥95%, Thermo Scientific™

Product Code. 10100732
Change view
Click to view available options
Quantity:
250 mg
1 g
25 g
Unit Size:
1g
250mg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10100732 1 g 1g
10601483 250 mg 250mg
10747285 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10100732 Supplier Thermo Scientific Maybridge Supplier No. CC18404DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 99662-34-7
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.187
MDL Number MFCD02681938
InChI Key PPGRDLZPSDHBIC-UHFFFAOYSA-N
Synonym 4-1h-pyrazol-1-yl benzaldehyde, 4-pyrazol-1-yl-benzaldehyde, 1-4'-formylphenyl pyrazole, 1-4-formylphenyl pyrazole, 4-1h-pyrazol-1-yl benzenecarbaldehyde, 4-pyrazol-1-yl benzaldehyde, 1-4-formylphenyl-1h-pyrazole, benzaldehyde, 4-1h-pyrazol-1-yl, 4-pyrazolylbenzaldehyde, pubchem22713
PubChem CID 2776477
IUPAC Name 4-pyrazol-1-ylbenzaldehyde
SMILES C1=CN(N=C1)C2=CC=C(C=C2)C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 172.19
Percent Purity 95%
Chemical Name or Material 4-(1H-Pyrazol-1-yl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.