missing translation for 'onlineSavingsMsg'
Learn More

4-(1,2,3-Thiadiazol-4-yl)benzamide, 95%, Thermo Scientific™

Product Code. 10141032
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10141032 10 g Amber Glass Bottle 10g
10324202 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10141032 Supplier Thermo Scientific Maybridge Supplier No. SEW01125EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175205-53-5
Molecular Formula C9H7N3OS
Molecular Weight (g/mol) 205.235
MDL Number MFCD00173809
InChI Key PAVNUEROKSNSDS-UHFFFAOYSA-N
Synonym 4-1,2,3-thiadiazol-4-yl benzamide, maybridge1_007554, 4-thiadiazol-4-yl benzamide, benzamide,4-1,2,3-thiadiazol-4-yl
PubChem CID 2739359
IUPAC Name 4-(thiadiazol-4-yl)benzamide
SMILES C1=CC(=CC=C1C2=CSN=N2)C(=O)N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 205.24
Percent Purity 95%
Chemical Name or Material 4-(1, 2, 3-Thiadiazol-4-yl)benzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.