missing translation for 'onlineSavingsMsg'
Learn More

[(3S,4R)-4-(4-Fluorophenyl)piperidin-3-yl]methanol (N-Desmethylparoxol), Mikromol™

Product Code. 17952886
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17952886 25 mg 25mg
17903476 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17952886 Supplier Mikromol Supplier No. MM1032.200025

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

[(3S,4R)-4-(4-Fluorophenyl)piperidin-3-yl]methanol (N-Desmethylparoxol)
TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material ((3S,4R)-4-(4-Fluorophenyl)piperidin-3-yl)methanol
CAS 125224-43-3
Analyte or Component Names ((3S,4R)-4-(4-Fluorophenyl)piperidin-3-yl)methanol
Shipping Condition Room Temperature
Molecular Formula C12 H16 F N O
Certifications/Compliance ISO 9001
Synonym [(3S,4R)-4-(4-Fluorophenyl)piperidin-3-yl]methanol, 3-Piperidinemethanol, 4-(4-fluorophenyl)-, (3S,4R)-, 3-Piperidinemethanol, 4-(4-fluorophenyl)-, (3S-trans)-, (3S,4R)-(-)-4-(4-Fluorophenyl)-3-piperidinemethanol, (3S,4R)-4-(4-Fluorophenyl)piperidine-3-methanol, Paroxetin Metabolite BRL 35961, Paroxetine Hydrochloride Impurity I, Paroxetine Impurity I
InChI Formula InChI=1S/C12H16FNO/c13-11-3-1-9(2-4-11)12-5-6-14-7-10(12)8-15/h1-4,10,12,14-15H,5-8H2/t10-,12-/m0/s1
SMILES OC[C@@H]1CNCC[C@H]1c2ccc(F)cc2
IUPAC Name ((3S,4R)-4-(4-Fluorophenyl)piperidin-3-yl)methanol
Molecular Weight (g/mol) 209.26
Quantity 25 mg
Formula Weight 209.1216 g/mol
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.