missing translation for 'onlineSavingsMsg'
Learn More

(3RS)-3-(2-Methoxy-5-methylphenyl)-N,N-bis(1-methylethyl)-3-phenylpropan-1-amine ((RS)-Tolterodine Methyl Ether), Mikromol™

Product Code. 17912926
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
17912926 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 17912926

Brand: Mikromol MM0900.080100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(3RS)-3-(2-Methoxy-5-methylphenyl)-N,N-bis(1-methylethyl)-3-phenylpropan-1-amine ((RS)-Tolterodine Methyl Ether)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3RS)-3-(2-Methoxy-5-methylphenyl)-N,N-bis(1-methylethyl)-3-phenylpropan-1-amine
CAS 124935-88-2
Analyte or Component Names (3RS)-3-(2-Methoxy-5-methylphenyl)-N,N-bis(1-methylethyl)-3-phenylpropan-1-amine
Shipping Condition Room Temperature
Molecular Formula C23 H33 N O
Certifications/Compliance ISO 9001
Synonym Benzenepropanamine, 2-methoxy-5-methyl-N,N-bis(1-methylethyl)-γ-phenyl-, 2-Methoxy-5-methyl-N,N-bis(1-methylethyl)-γ-phenylbenzenepropanamine, N,N-Diisopropyl-3-(2-methoxy-5-methylphenyl)-3-phenylpropylamine, N-[3-(2-Methoxy-5-methylphenyl)-3-phenylpropyl]-diisopropylamine, (3RS)-3-(2-Methoxy-5-methylphenyl)-N,N-bis(1-methylethyl)-3-phenylpropan-1-amine, Tolterodine Tartrate Imp. C (EP), Tolterodine Tartrate Impurity C, Tolterodine Impurity C
InChI Formula InChI=1S/C23H33NO/c1-17(2)24(18(3)4)15-14-21(20-10-8-7-9-11-20)22-16-19(5)12-13-23(22)25-6/h7-13,16-18,21H,14-15H2,1-6H3
SMILES COc1ccc(C)cc1C(CCN(C(C)C)C(C)C)c2ccccc2
IUPAC Name 3-(2-methoxy-5-methylphenyl)-3-phenyl-N,N-di(propan-2-yl)propan-1-amine
Molecular Weight (g/mol) 339.5142
Quantity 100 mg
Formula Weight 339.2562 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.