missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl Hippuric Acid-d7, TRC

Product Code. 30466394
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30466394 10 mg 10mg
30469837 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30466394 Supplier Toronto Research Chemicals Supplier No. TRCM31198710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1216551-07-3
Molecular Formula C10 D7 H4 N O3
Molecular Weight (g/mol) 200.242
Synonym 2-[[2,3,4,6-Tetradeuterio-5-(trideuteriomethyl)benzoyl]amino]acetic acid, N-(3-Methylbenzoyl)glycine-D7 (methylbenzoyl-D7), N-(3-Methylbenzoyl)glycine-d7, 3-Methylhippuric acid-d7, 2-[[3-(Methyl-d3)benzoyl-2,4,5,6-d4]amino]acetic acid, m-Methylhippuric acid-d7, m-Toluric acid-d7, 3-Methyl hippuric acid-d7
IUPAC Name 2-[[2,3,4,6-tetradeuterio-5-(trideuteriomethyl)benzoyl]amino]acetic acid
SMILES [2H]c1c([2H])c(C(=O)NCC(=O)O)c([2H])c(c1[2H])C([2H])([2H])[2H]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H11NO3/c1-7-3-2-4-8(5-7)10(14)11-6-9(12)13/h2-5H,6H2,1H3,(H,11,14)(H,12,13)/i1D3,2D,3D,4D,5D
Formula Weight 200.118
Percent Purity >95%
Chemical Name or Material 3-Methyl Hippuric Acid-d7
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.