missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-phenyl-4-isoxazolecarboxylic acid, 97%, Thermo Scientific™

Product Code. 10153434
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10153434 10 g Amber Glass Bottle 10g
10213022 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10153434 Supplier Thermo Scientific Maybridge Supplier No. CC20301EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 17153-21-8
Molecular Formula C11H9NO3
Molecular Weight (g/mol) 203.197
InChI Key GLNQCTGGLIXRRJ-UHFFFAOYSA-N
Synonym 3-methyl-5-phenylisoxazole-4-carboxylic acid, 3-methyl-5-phenyl-4-isoxazolecarboxylic acid, 4-carboxy-3-methylisoxazol-5-yl benzene, 3-methyl-5-phenyl-4-isoxazole carboxylic acid, 3-methyl-5-phenyl-isoxazole-4-carboxylic acid, 4-isoxazolecarboxylicacid, 3-methyl-5-phenyl, 4-isoxazolecarboxylic acid, 3-methyl-5-phenyl
PubChem CID 2776514
IUPAC Name 3-methyl-5-phenyl-1,2-oxazole-4-carboxylic acid
SMILES CC1=NOC(=C1C(=O)O)C2=CC=CC=C2

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 203.2
Percent Purity 97%
Chemical Name or Material 3-Methyl-5-phenyl-4-isoxazolecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.