missing translation for 'onlineSavingsMsg'
Learn More

(3-Glycidoxypropyl)trimethoxysilane, 97%

Product Code. p-7024164
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438357 25 g 25g
11448357 100 g 100g
11458357 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438357 Supplier Thermo Scientific Alfa Aesar Supplier No. A18431.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 2530-83-8 | C9H20O5Si | 236.339 g/mol

(3-Glycidoxypropyl)trimethoxysilane is used in the production of carbon steel and its pre treatment favors the dry and wet adhesion of epoxy coating. It is also used to prepare epoxy-functionalized silica nanoparticles, which gives a reactive surface for one-step and high-density protein immobilization. It serves as a coupling agent and adhesion promoter.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Glycidoxypropyl)trimethoxysilane is used in the production of carbon steel and its pre treatment favors the dry and wet adhesion of epoxy coating. It is also used to prepare epoxy-functionalized silica nanoparticles, which gives a reactive surface for one-step and high-density protein immobilization. It serves as a coupling agent and adhesion promoter.

Solubility
Miscible with alcohols, ketones and aliphatic or aromatic hydrocarbons. Immiscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2530-83-8
Molecular Formula C9H20O5Si
Molecular Weight (g/mol) 236.339
MDL Number MFCD00005144
InChI Key BPSIOYPQMFLKFR-UHFFFAOYSA-N
Synonym 3-glycidoxypropyltrimethoxysilane, 3-glycidoxypropyl trimethoxysilane, glymo, silicone kbm 403, silane a 187, union carbide a-187, silan a 187, silane z 6040, silane-y-4087, 3-glycidyloxypropyltrimethoxysilane
PubChem CID 17317
IUPAC Name trimethoxy-[3-(oxiran-2-ylmethoxy)propyl]silane
SMILES CO[Si](CCCOCC1CO1)(OC)OC

Specifications

Boiling Point 119°C to 121°C (2mmHg)
Melting Point <-70°C
Quantity 25 g
Beilstein 4308125
Flash Point 135°C (275°F)
Solubility Information Miscible with alcohols,ketones and aliphatic or aromatic hydrocarbons. Immiscible with water.
Formula Weight 236.34
Percent Purity 97%
Odor Odorless
Refractive Index 1.429
Sensitivity Moisture sensitive
Density 1.072
Chemical Name or Material (3-Glycidoxypropyl)trimethoxysilane
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.