missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)-5-phenyl-1,2,4-oxadiazole, 97%, Thermo Scientific™

Product Code. 10130702
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10130702 1 g 1g
10089581 250 mg 250mg
10168414 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10130702 Supplier Thermo Scientific Maybridge Supplier No. SEW02030DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1201-68-9
Molecular Formula C9H7ClN2O
Molecular Weight (g/mol) 194.618
MDL Number MFCD00084968
InChI Key VEIXFEJMHJPUBS-UHFFFAOYSA-N
PubChem CID 302154
IUPAC Name 3-(chloromethyl)-5-phenyl-1,2,4-oxadiazole
SMILES C1=CC=C(C=C1)C2=NC(=NO2)CCl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 3-chloromethyl-5-phenyl-1,2,4-oxadiazole, 3-chloromethyl-5-phenyl-1,2,4 oxadiazole, 1,2,4-oxadiazole, 3-chloromethyl-5-phenyl, 3-chloromethyl-5-phenyl-1,2,4-oxadiazol, 1,2,4-oxadiazole,3-chloromethyl-5-phenyl
Formula Weight 194.62
Percent Purity 97%
Chemical Name or Material 3-(Chloromethyl)-5-phenyl-1, 2, 4-oxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.