missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzoyl chloride, 99+%

Product Code. 10020840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
50 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
50mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10020840 100 mL Glass bottle 100mL
10172610 5 mL Glass bottle 5mL
10010840 50 mL Glass bottle 50mL
10030850 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10020840 Supplier Thermo Scientific Acros Supplier No. 108741000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 618-46-2 | C7H4Cl2O | 175.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-46-2
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.01
MDL Number MFCD00000671
InChI Key WHIHIKVIWVIIER-UHFFFAOYSA-N
Synonym m-chlorobenzoyl chloride, benzoyl chloride, 3-chloro, benzoyl chloride, m-chloro, m-chlorobenzoylchloride, 3-chlorobenzoylchloride, benzoylchloride, 3-chloro, mcoc, 3-chlorobenzoic acid chloride, ccris 8615, 3chlorobenzoyl chloride
PubChem CID 69252
IUPAC Name 3-chlorobenzoyl chloride
SMILES ClC(=O)C1=CC=CC(Cl)=C1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Colorless
Density 1.3600g/mL
Boiling Point 223.0°C to 225.0°C
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 0.03% max. Total other poly-halogenated acid halides (GC)
Packaging Glass bottle
Linear Formula ClC6H4COCl
Refractive Index 1.5680 to 1.5700
Quantity 100 mL
Beilstein 09, 338
Specific Gravity 1.36
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in alcohol,ether and acetone
Formula Weight 175.01
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 3-Chlorobenzoyl chloride, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Chlorobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.