missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-fluorobenzenesulfonyl chloride, 98%

Product Code. 11483544
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483544 5 g 5g
11493544 25 g 25g
11403554 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483544 Supplier Thermo Scientific Alfa Aesar Supplier No. A11222.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91170-93-3 | C6H3Cl2FO2S | 229.046 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91170-93-3
Molecular Formula C6H3Cl2FO2S
Molecular Weight (g/mol) 229.046
MDL Number MFCD00041444
InChI Key DXFXNSNBZNELII-UHFFFAOYSA-N
Synonym 3-chloro-4-fluorobenzene-1-sulfonyl chloride, 3-chloro-4-fluorobenzenesulphonyl chloride, 3-chloro-4-fluorobenzenesulfonylchloride, benzenesulfonyl chloride, 3-chloro-4-fluoro, 3-chloro-4-fluorophenylsulfonyl chloride, 3-chloro-4-fluoro-benzenesulfonyl chloride, benzenesulfonyl chloride, 3-chloro-4-fluoro-9ci, pubchem5093, acmc-209r8d, ksc488g2p
PubChem CID 145222
IUPAC Name 3-chloro-4-fluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1S(=O)(=O)Cl)Cl)F

Specifications

Density 1.61
Boiling Point 104°C to 105°C (1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.552
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 229.06
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-fluorobenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.