missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromoacetyl)coumarin, 97%, Thermo Scientific™

Product Code. p-7077623 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15422172 5 g 5g
15432172 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15422172 Supplier Thermo Scientific Acros Supplier No. 456550050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 29310-88-1
Molecular Formula C11H7BrO3
Molecular Weight (g/mol) 267.08
MDL Number MFCD00488331
InChI Key NTYOLVNSXVYRTJ-UHFFFAOYSA-N
Synonym 3-bromoacetyl coumarin, 3-2-bromoacetyl-2h-chromen-2-one, 3-2-bromoacetyl chromen-2-one, 3-bromoacetylcoumarin, 3-bromoacetyl-2h-chromen-2-one, 2h-1-benzopyran-2-one, 3-bromoacetyl, pubchem8670, rarechem ab ka k003, 3-2-bromoacetyl-coumarin
PubChem CID 2063461
IUPAC Name 3-(2-bromoacetyl)chromen-2-one
SMILES BrCC(=O)C1=CC2=CC=CC=C2OC1=O

Specifications

Melting Point 164.0°C to 168.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 267.08
Percent Purity 97%
Chemical Name or Material 3-(Bromoacetyl)coumarin
compliance-icons compliance-icons
Product Identifier
  • DEUX 3-(Bromoacetyl)coumarin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.