missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-naphthoic acid, 85%, tech.

Product Code. 10071841 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10071841 5 g Glass bottle 5g
10001871 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10071841 Supplier Thermo Scientific Acros Supplier No. 104140050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5959-52-4 | C11H9NO2 | 187.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5959-52-4
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.2
MDL Number MFCD00004115
InChI Key XFXOLBNQYFRSLQ-UHFFFAOYSA-N
Synonym 3-amino-2-naphthoic acid, 2-amino-3-naphthoic acid, 3-aminoisonaphthoic acid, 2-naphthalenecarboxylic acid, 3-amino, 2-amino-3-carboxynaphthalene, 2-naphthoic acid, 3-amino, 3-amino-2-naphthalenecarboxylic acid, unii-iv5z0phl5d, iv5z0phl5d, 3-amino-naphthalene-2-carboxylic acid
PubChem CID 22244
IUPAC Name 3-aminonaphthalene-2-carboxylic acid
SMILES C1=CC=C2C=C(C(=CC2=C1)C(=O)O)N

Specifications

CAS Min % 80.0
CAS Max % 100.0
Melting Point 210.0°C
Color Green to Green-Yellow
Infrared Spectrum Authentic
Assay Percent Range 85%
Packaging Glass bottle
Linear Formula H2NC10H6CO2H
Quantity 5 g
Beilstein 14, 535
Merck Index 15, 448
Formula Weight 187.2
Percent Purity 85%
Grade Technical
Physical Form Powder
Chemical Name or Material 3-Amino-2-naphthoic acid, 85%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Amino-2-naphthoic acid, tech.
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.