missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethyl-1H-pyrazole, 99%

Product Code. p-7080577
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489432 50 g 50g
11499432 250 g 250g
11479432 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489432 Supplier Thermo Scientific Alfa Aesar Supplier No. A10157.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 67-51-6 | C5H8N2 | 96.13 g/mol

3,5-Dimethyl-1H-pyrazole is combined with Chromium(VI)­ oxide, is a valuable reagent for oxidation of primary and secondary alcohols to carbonyl compounds. It reacts with malonic esters gives a family of cross-conjugated monomeric betaines. It is a common reagent for the preparation of pyrazolato ligated complexes. It is also used to prepare N-1-substituted derivatives having antibacterial activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethyl-1H-pyrazole is combined with Chromium(VI)­ oxide, is a valuable reagent for oxidation of primary and secondary alcohols to carbonyl compounds. It reacts with malonic esters gives a family of cross-conjugated monomeric betaines. It is a common reagent for the preparation of pyrazolato ligated complexes. It is also used to prepare N-1-substituted derivatives having antibacterial activity.

Solubility
Soluble in water, methanol 0.1 g/mL.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-51-6
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.13
MDL Number MFCD00005243
InChI Key SDXAWLJRERMRKF-UHFFFAOYSA-N
Synonym 3,5-dimethylpyrazole, 1h-pyrazole, 3,5-dimethyl, 3,5 dmp, pyrazole, 3,5-dimethyl, 3,5-dwumetylopirazolu, unii-h21n865k9j, 3,5-dwumetylopirazolu polish, 3,5-dimethyl pyrazole, pyrazole,5-dimethyl, pubchem13256
PubChem CID 6210
IUPAC Name 3,5-dimethyl-1H-pyrazole
SMILES CC1=CC(C)=NN1

Specifications

Melting Point 105°C to 108°C
pH 6.7
Boiling Point 218°C
Quantity 50 g
Beilstein 106325
Solubility Information Soluble in water,methanol 0.1g/mL.
Formula Weight 96.13
Percent Purity 99%
Chemical Name or Material 3,5-Dimethyl-1H-pyrazole
compliance-icons compliance-icons
Product Identifier
  • 3,5-Dimethyl-1H-pyrazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 3,5-Dimethyl-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.