missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butylaniline, 97%, Thermo Scientific Chemicals

Product Code. 15465527
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15465527 250 mg 250mg
15455527 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465527 Supplier Thermo Scientific Alfa Aesar Supplier No. H32768.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2380-36-1
Molecular Formula C14H23N
Molecular Weight (g/mol) 205.345
MDL Number MFCD00134096
InChI Key MJKNHXCPGXUEDO-UHFFFAOYSA-N
Synonym 3,5-di-tert-butylaniline, 3,5-di-t-butylaniline, benzenamine, 3,5-bis 1,1-dimethylethyl, acmc-1cdyp, 3,5-ditert-butylphenylamine, 3,5-bis tert-butyl aniline, 3,5-di-tert-butylphenylamine, 3,5-bis tert-butyl phenylamine, mjknhxcpgxuedo-uhfffaoysa, 3,5-ditert-butyl-phenyl-amine
PubChem CID 75419
IUPAC Name 3,5-ditert-butylaniline
SMILES CC(C)(C)C1=CC(=CC(=C1)N)C(C)(C)C

Specifications

Melting Point 54°C to 57°C
Quantity 250 mg
Formula Weight 205.35
Percent Purity 97%
Chemical Name or Material 3,5-Di-tert-butylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.