Learn More
3-(4-Morpholinyl)propylamine, 98%
Description
3-(4-Morpholinyl)propylamine is catalytic agent, petrochemical additive. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-(4-Morpholinyl)propylamine is catalytic agent, petrochemical additive. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Fully miscible in water.
Notes
Air Sensitive. Store under dry inert gas. Store away from air. Incompatible with acids, acid chlorides, acid anhydrides.
Specifications
Specifications
| Melting Point | -15°C |
| Density | 0.987 |
| Boiling Point | 223°C to 225°C |
| Flash Point | 98°C (210°F) |
| Refractive Index | 1.4761 |
| Quantity | 250 g |
| UN Number | UN2735 |
| Sensitivity | Air Sensitive |
| Solubility Information | Fully miscible in water. |
| Formula Weight | 144.22 |
| Show More |
Safety and Handling
- 3-(4-Morpholinyl)propylamine
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: N-Aminopropylmorpholine
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.